Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031689.1 | NP_113877.1 | 5 | 450 | ||
GenBank mRNA | AF013247.1 | - | 5 | 450 |