Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097589.1 | NP_001091058.1 | 2 | 712 | ||
NM_005416.2 | NP_005407.1 | 3 | 796 | |||
GenBank mRNA | AJ243667.1 | - | 3 | 670 | ||
AY118269.1 | - | 2 | 688 | |||
BC017802.1 | - | 3 | 686 | 4288753 | ||
EF553525.1 | - | 3 | 1736 |