Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003126.2 | NP_003117.2 | 5 | 782 | ||
XM_011509916.2 | XP_011508218.1 | 6 | 660 | |||
XM_011509917.2 | XP_011508219.1 | 5 | 696 | |||
XM_011509918.2 | XP_011508220.1 | 5 | 696 | |||
XM_011509919.2 | XP_011508221.1 | 5 | 698 | |||
GenBank mRNA | M61877.1 | - | 5 | 769 |