Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pllp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022533.1 | NP_071978.1 | 2 | 485 | ||
GenBank mRNA | BC078823.1 | - | 2 | 365 | 7108239 | |
U13617.1 | - | 2 | 258 | |||
Z49858.1 | - | 2 | 485 |