Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172067.1 | NP_742064.1 | 5 | 856 | ||
GenBank mRNA | BC107655.1 | - | 5 | 933 | 7935103 | |
M88469.1 | - | 5 | 856 |