Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171994.4 | NP_741991.3 | 4 | 278 | ||
XM_008764286.1 | XP_008762508.1 | 4 | 783 | |||
XM_008764287.1 | XP_008762509.1 | 4 | 358 | |||
GenBank mRNA | AF491841.1 | - | 3 | 271 | ||
BC060535.1 | - | 4 | 278 | 6920523 | ||
FQ213007.1 | - | 4 | 277 | |||
FQ225045.1 | - | 4 | 277 | |||
FQ225541.1 | - | 4 | 390 | |||
FQ225902.1 | - | 4 | 277 | |||
FQ225973.1 | - | 4 | 390 | |||
FQ226943.1 | - | 4 | 273 | |||
FQ227167.1 | - | 4 | 277 | |||
FQ227499.1 | - | 4 | 277 | |||
FQ227894.1 | - | 4 | 277 | |||
FQ228149.1 | - | 4 | 390 | |||
FQ230725.1 | - | 4 | 286 | |||
FQ231089.1 | - | 4 | 277 | |||
FQ231482.1 | - | 4 | 286 | |||
FQ231846.1 | - | 4 | 390 | |||
FQ232179.1 | - | 4 | 277 | |||
FQ232295.1 | - | 4 | 286 | |||
FQ232459.1 | - | 4 | 277 | |||
FQ233586.1 | - | 4 | 288 | |||
FQ234217.1 | - | 4 | 277 | |||
FQ234639.1 | - | 4 | 286 | |||
FQ234830.1 | - | 4 | 286 |