Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172066.1 | NP_742063.1 | 1 | 398 | ||
GenBank mRNA | BC086981.1 | - | 1 | 364 | 7112403 | |
Y16774.1 | - | 1 | 398 |