Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172072.2 | NP_742069.1 | 14 | 1611 | ||
GenBank mRNA | BC081705.1 | - | 14 | 1611 | 7192410 | |
U81160.1 | - | 14 | 1611 |