Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022588.1 | NP_072110.1 | 13 | 1356 | ||
XM_008764949.1 | XP_008763171.1 | 13 | 1356 | |||
XM_008764950.1 | XP_008763172.1 | 13 | 1356 | |||
XM_008764951.1 | XP_008763173.1 | 13 | 1355 | |||
XM_008764952.1 | XP_008763174.1 | 12 | 1305 | |||
XM_008764953.1 | XP_008763175.1 | 13 | 1355 | |||
XM_008764954.1 | XP_008763176.1 | 13 | 1355 | |||
XM_008764955.1 | XP_008763177.1 | 12 | 1304 | |||
XM_008764956.1 | XP_008763178.1 | 12 | 1304 | |||
XM_008764957.1 | XP_008763179.1 | 12 | 1304 | |||
XM_008764958.1 | XP_008763180.1 | 13 | 1309 | |||
GenBank mRNA | U02522.1 | - | 13 | 1356 |