Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synj2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022599.2 | NP_072121.2 | 2 | 170 | ||
GenBank mRNA | AF260258.1 | - | 2 | 170 | ||
BC126067.1 | - | 2 | 158 | 7323715 |