Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256891.1 | NP_001243820.1 | 6 | 484 | ||
NM_001256892.1 | NP_001243821.1 | 6 | 460 | |||
NM_003130.3 | NP_003121.1 | 6 | 484 | |||
NM_198901.1 | NP_944490.1 | 6 | 460 | |||
XM_011516528.1 | XP_011514830.1 | 5 | 354 | |||
GenBank mRNA | AK129770.1 | - | 6 | 453 | ||
AK296601.1 | - | 6 | 443 | |||
AK310366.1 | - | 6 | 460 | |||
BC011025.1 | - | 6 | 442 | 4281626 | ||
BI559592.1 | - | 6 | 478 | |||
BI597920.1 | - | 6 | 480 | |||
BM739144.1 | - | 6 | 460 | |||
BM793972.1 | - | 3 | 202 | |||
BU854353.1 | - | 2 | 123 | |||
DA268050.1 | - | 6 | 484 | |||
L12387.1 | - | 6 | 430 | |||
M32886.1 | - | 6 | 438 |