Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022624.1 | NP_072146.1 | 11 | 2558 | ||
XM_006253845.2 | XP_006253907.1 | 10 | 1814 | |||
GenBank mRNA | AF184921.1 | - | 7 | 802 | ||
BC158843.1 | - | 10 | 1433 |