Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022681.2 | NP_073172.2 | 4 | 2321 | ||
GenBank mRNA | AF234680.1 | - | 1 | 1341 |