Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022686.2 | NP_073177.1 | 2 | 769 | ||
GenBank mRNA | AY936209.1 | - | 2 | 769 | ||
BC100619.1 | - | 2 | 773 | 7938313 |