Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022699.3 | NP_073190.1 | 3, 4 | 246 | ||
GenBank mRNA | BC058471.1 | - | 3, 4 | 250 | 6919868 | |
FQ210232.1 | - | 3, 4 | 243 | |||
FQ210454.1 | - | 3, 4 | 242 | |||
FQ210629.1 | - | 3, 4 | 246 | |||
FQ211005.1 | - | 3, 4 | 248 | |||
FQ211389.1 | - | 3, 4 | 244 | |||
FQ211694.1 | - | 3, 4 | 243 | |||
FQ211731.1 | - | 3, 4 | 242 | |||
FQ211953.1 | - | 3, 4 | 246 | |||
FQ214466.1 | - | 3, 4 | 243 | |||
FQ217830.1 | - | 3, 4 | 243 | |||
FQ217913.1 | - | 3, 4 | 243 | |||
FQ221046.1 | - | 3, 4 | 243 | |||
FQ221166.1 | - | 3, 4 | 243 | |||
FQ221653.1 | - | 3, 4 | 251 | |||
FQ221662.1 | - | 3, 4 | 244 | |||
FQ221710.1 | - | 3, 4 | 243 | |||
FQ221740.1 | - | 3, 4 | 242 | |||
FQ221836.1 | - | 3, 4 | 243 | |||
FQ222551.1 | - | 3, 4 | 243 | |||
FQ222688.1 | - | 3, 4 | 242 | |||
FQ222703.1 | - | 3, 4 | 242 | |||
FQ223191.1 | - | 3, 4 | 242 | |||
FQ223319.1 | - | 3, 4 | 243 | |||
FQ223321.1 | - | 3, 4 | 224 | |||
FQ223373.1 | - | 3, 4 | 242 | |||
FQ223410.1 | - | 3, 4 | 340 | |||
FQ223604.1 | - | 563 | ||||
FQ223661.1 | - | 3, 4 | 243 | |||
FQ224433.1 | - | 3, 4 | 245 | |||
FQ224506.1 | - | 3, 4 | 242 | |||
FQ224779.1 | - | 3, 4 | 243 | |||
FQ228655.1 | - | 3, 4 | 242 | |||
FQ228991.1 | - | 3, 4 | 242 | |||
FQ229525.1 | - | 3, 4 | 241 | |||
K02932.1 | - | 2, 3 | 173 |
Gene Symbol: LOC100362027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727709.3 | XP_002727755.1 | 1 | 250 |
Gene Symbol: LOC103690975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758743.1 | XP_008756965.1 | 2 | 287 |
Gene Symbol: RGD1562394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_344225.3 | XP_344226.1 | 1 | 184 |
Gene Symbol: Rpl30l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_344178.3 | XP_344179.2 | 1 | 173 |