Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022704.2 | NP_073195.2 | 4 | 608 | ||
XM_006231249.2 | XP_006231311.1 | 6 | 884 | |||
XM_006231250.2 | XP_006231312.1 | 6 | 804 | |||
GenBank mRNA | AY325174.1 | - | 638 | |||
AY325178.1 | - | 638 | ||||
FQ210356.1 | - | 6 | 793 | |||
M14103.1 | - | 6 | 778 | |||
X05023.1 | - | 4 | 608 |
Gene Symbol: LOC100911854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749087.3 | XP_003749135.1 | 884 | |||
XM_003749088.3 | XP_003749136.1 | 804 |