Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022712.1 | NP_073203.1 | 18 | 2160 | ||
XM_006248462.2 | XP_006248524.1 | 18 | 2270 | |||
XM_006248463.2 | XP_006248525.1 | 18 | 2182 | |||
GenBank mRNA | CV111368.1 | - | 1 | 127 | ||
M58040.1 | - | 15 | 1611 |