Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022939.2 | NP_075228.2 | 8, 9 | 804 | ||
GenBank mRNA | AF035632.1 | - | 8, 9 | 720 |