Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inppl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270843.1 | NP_001257772.1 | 18 | 2345 | ||
NM_022944.2 | NP_075233.1 | 19 | 2401 | |||
GenBank mRNA | AB011439.1 | - | 18 | 2345 | ||
AB025794.1 | - | 19 | 2399 | |||
CK653108.1 | - | 2 | 127 |