Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003141.3 | NP_003132.2 | 7 | 1086 | ||
GenBank mRNA | AK225564.1 | - | 5 | 799 | ||
AY742713.1 | - | 6 | 973 | |||
BC010861.1 | - | 7 | 1057 | 3900540 | ||
M34551.1 | - | 6 | 1011 | |||
M62800.1 | - | 7 | 1058 |