Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031703.1 | NP_113891.1 | 3 | 377 | ||
GenBank mRNA | BC127490.1 | - | 3 | 359 | 8366493 | |
D17695.1 | - | 3 | 292 | |||
L35108.1 | - | 3 | 377 |