Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031711.1 | NP_113899.1 | 4 | 432 | ||
XM_006230899.1 | XP_006230961.1 | 4 | 559 | |||
GenBank mRNA | Y12708.1 | - | 4 | 432 |