Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031721.1 | NP_113909.1 | 7 | 1188 | ||
GenBank mRNA | AF179370.1 | - | 7 | 1188 | ||
BC081767.1 | - | 7 | 1213 | 7121959 | ||
FQ220294.1 | - | 4 | 279 | |||
FQ228889.1 | - | 6 | 640 | |||
FQ228942.1 | - | 6 | 666 | |||
FQ229223.1 | - | 2 | 79 | |||
FQ229948.1 | - | 4 | 337 |
Gene Symbol: Psmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ228875.1 | - | 58 |