Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031724.1 | NP_113912.1 | 7 | 839 | ||
XM_006232753.2 | XP_006232815.1 | 7 | 1294 | |||
XM_006232754.2 | XP_006232816.1 | 7 | 1344 | |||
GenBank mRNA | AF295404.1 | - | 4 | 358 | ||
DV726194.1 | - | 4 | 380 |