Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031724.1 | NP_113912.1 | 1 | 162 | ||
XM_006232753.2 | XP_006232815.1 | 1 | 620 | |||
XM_006232754.2 | XP_006232816.1 | 1 | 620 | |||
GenBank mRNA | CK599635.1 | - | 1 | 162 |