Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256510.1 | NP_001243439.1 | 6, 7 | 832 | ||
NM_001256511.1 | NP_001243440.1 | 6, 7 | 713 | |||
NM_001256512.1 | NP_001243441.1 | 6, 7 | 695 | |||
NM_001256513.1 | NP_001243442.1 | 6, 7 | 623 | |||
NM_003143.2 | NP_003134.1 | 6, 7 | 530 | |||
NR_046269.1 | 7, 8 | 580 | ||||
GenBank mRNA | AF277319.1 | - | 6, 7 | 476 | ||
AK313033.1 | - | 6, 7 | 543 | |||
BC000895.1 | - | 6, 7 | 832 | 3464282 | ||
BC093054.1 | - | 6, 7 | 492 | 30722138 | ||
BF131408.1 | - | 7, 8 | 511 | |||
BG030060.1 | - | 646 | ||||
BP308841.1 | - | 5, 6 | 399 | |||
BQ959702.1 | - | 6, 7 | 595 | |||
M94556.1 | - | 6, 7 | 475 |