Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031729.1 | NP_113917.1 | 6 | 785 | ||
XM_006228465.1 | XP_006228527.1 | 5 | 1316 | |||
GenBank mRNA | BC078786.1 | - | 6 | 812 | 7126555 | |
U12203.1 | - | 6 | 713 | |||
X77237.1 | - | 6 | 785 |