Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031745.2 | NP_113933.2 | 3 | 552 | ||
XM_006249354.2 | XP_006249416.1 | 3 | 552 | |||
XM_008769250.1 | XP_008767472.1 | 3 | 517 | |||
XM_008769251.1 | XP_008767473.1 | 2 | 478 | |||
XM_008769252.1 | XP_008767474.1 | 2 | 478 | |||
XM_008769253.1 | XP_008767475.1 | 2 | 478 | |||
XM_008769254.1 | XP_008767476.1 | 2 | 478 | |||
XM_008769256.1 | XP_008767478.1 | 2 | 478 | |||
XM_008769257.1 | XP_008767479.1 | 2 | 478 | |||
XM_008769258.1 | XP_008767480.1 | 2 | 478 | |||
XM_008769259.1 | XP_008767481.1 | 2 | 478 | |||
GenBank mRNA | AJ237670.1 | - | 3 | 552 | ||
BC097264.1 | - | 3 | 500 |