Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031745.2 | NP_113933.2 | 18 | 3378 | ||
XM_006249354.2 | XP_006249416.1 | 18 | 3378 | |||
XM_008769250.1 | XP_008767472.1 | 21 | 5989 | |||
XM_008769251.1 | XP_008767473.1 | 20 | 5950 | |||
XM_008769252.1 | XP_008767474.1 | 19 | 5917 | |||
XM_008769253.1 | XP_008767475.1 | 19 | 5845 | |||
XM_008769254.1 | XP_008767476.1 | 18 | 5812 | |||
XM_008769255.1 | XP_008767477.1 | 12 | 7092 | |||
XM_008769256.1 | XP_008767478.1 | 19 | 3682 | |||
XM_008769257.1 | XP_008767479.1 | 19 | 3682 | |||
XM_008769258.1 | XP_008767480.1 | 18 | 3649 | |||
XM_008769259.1 | XP_008767481.1 | 17 | 3544 | |||
GenBank mRNA | AJ237670.1 | - | 18 | 3378 |