Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023027.1 | NP_075416.1 | 3 | 193 | ||
GenBank mRNA | AF181856.1 | - | 3 | 193 |