Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308197.1 | NP_001295126.1 | 4 | 533 | ||
NM_001308204.1 | NP_001295133.1 | 4 | 359 | |||
NM_001308205.1 | NP_001295134.1 | 4 | 383 | |||
NM_007107.4 | NP_009038.1 | 4 | 494 | |||
GenBank mRNA | AF087907.1 | - | 4 | 442 | ||
AF110647.1 | - | 4 | 439 | |||
AK304364.1 | - | 4 | 431 | |||
AK312935.1 | - | 4 | 401 | |||
BC017203.1 | - | 4 | 440 | 3618589 | ||
BT006669.1 | - | 4 | 383 | |||
DA135338.1 | - | 4 | 478 | |||
DA283706.1 | - | 4 | 359 | |||
DA725890.1 | - | 4 | 385 | |||
DA750309.1 | - | 4 | 478 |