Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023960.2 | NP_076450.1 | 1 | 277 | ||
GenBank mRNA | AB050637.1 | - | 1 | 259 | ||
AY028605.1 | - | 1 | 277 | |||
BC161829.1 | - | 1 | 457 | 9033053 |