Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023965.1 | NP_076455.1 | 6, 7 | 671 | ||
GenBank mRNA | AJ295950.1 | - | 6, 7 | 671 | ||
FQ217794.1 | - | 740 | ||||
FQ231233.1 | - | 723 |