Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031749.2 | NP_113937.1 | 2 | 563 | ||
GenBank mRNA | AF087431.1 | - | 2 | 564 |