Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023979.1 | NP_076469.1 | 16 | 2838 | ||
XM_008765263.1 | XP_008763485.1 | 13 | 2630 | |||
GenBank mRNA | AF218388.1 | - | 16 | 2293 | ||
AF320222.1 | - | 16 | 2838 |