Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023989.1 | NP_076479.1 | 8, 9 | 844 | ||
XM_008768805.1 | XP_008767027.1 | 8, 9 | 975 | |||
GenBank mRNA | AF260129.1 | - | 8, 9 | 844 | ||
FQ212134.1 | - | 8, 9 | 939 | |||
FQ222500.1 | - | 7, 8 | 787 |