Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278697.1 | NP_001265626.1 | 3 | 306 | ||
NM_003147.5 | NP_003138.3 | 3 | 306 | |||
NM_175698.2 | NP_783629.1 | 3 | 306 | |||
GenBank mRNA | BC002818.2 | - | 3 | 240 | 3636268 | |
BC007343.2 | - | 3 | 240 | 3615133 | ||
BC016957.1 | - | 3 | 240 | 3916128 | ||
BC069313.1 | - | 2 | 170 | 7262129 | ||
BC071827.1 | - | 3 | 240 | 4641362 | ||
BC103863.2 | - | 3 | 308 | 40003337 | ||
BG104299.1 | - | 3 | 278 | |||
BI560469.1 | - | 3 | 313 | |||
JX014236.1 | - | 2 | 170 | |||
X86175.1 | - | 3 | 261 | |||
Z49105.1 | - | 3 | 268 |
Gene Symbol: SSX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164417.2 | NP_001157889.1 | 3 | 305 | ||
NM_001278701.1 | NP_001265630.1 | 3 | 305 | |||
NM_001278702.1 | NP_001265631.1 | 3 | 305 | |||
GenBank mRNA | BE391023.1 | - | 3 | 259 | ||
CB158955.1 | - | 3 | 222 |