Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024374.4 | NP_077350.1 | 2, 3 | 304 | ||
GenBank mRNA | AY951952.1 | - | 311 | |||
BC088136.1 | - | 2, 3 | 304 | 7377576 | ||
D26179.1 | - | 2, 3 | 359 | |||
FQ228349.1 | - | 425 | ||||
U21661.1 | - | 2, 3 | 383 |