Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024377.1 | NP_077353.1 | 3 | 1112 | ||
GenBank mRNA | M95780.1 | - | 3 | 1112 |