Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007559.2 | NP_001007560.1 | 5 | 501 | ||
NM_001308201.1 | NP_001295130.1 | 5 | 476 | |||
NM_005637.3 | NP_005628.2 | 5 | 501 | |||
XM_006722527.2 | XP_006722590.1 | 6 | 527 | |||
XM_011526145.1 | XP_011524447.1 | 5 | 469 | |||
XM_011526147.2 | XP_011524449.1 | 7 | 752 | |||
XM_011526148.2 | XP_011524450.1 | 7 | 739 | |||
XM_011526149.2 | XP_011524451.1 | 7 | 764 | |||
XM_011526150.2 | XP_011524452.1 | 6 | 624 | |||
XM_011526151.2 | XP_011524453.1 | 7 | 753 | |||
XM_011526152.2 | XP_011524454.1 | 6 | 624 | |||
GenBank mRNA | AF244972.1 | - | 5 | 438 | ||
AF343880.1 | - | 5 | 435 | |||
AK289510.1 | - | 5 | 479 | |||
AK296949.1 | - | 5 | 459 | |||
AK299082.1 | - | 4 | 369 | |||
AK301504.1 | - | 5 | 506 | |||
AK304305.1 | - | 6 | 617 | |||
AK307923.1 | - | 4 | 392 | |||
AK316302.1 | - | 5 | 451 | |||
AU279615.1 | - | 5 | 506 | |||
BC096221.1 | - | 5 | 438 | 40004908 | ||
BC096222.1 | - | 5 | 438 | 40004909 | ||
BC096223.1 | - | 5 | 438 | 40004911 | ||
BC096224.1 | - | 5 | 438 | 40004912 | ||
CR542088.1 | - | 5 | 435 | |||
CR542103.1 | - | 5 | 435 | |||
DC378335.1 | - | 5 | 476 | |||
X79201.1 | - | 5 | 438 |