Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024384.2 | NP_077360.2 | 9 | 861 | ||
GenBank mRNA | AF275266.1 | - | 2 | 126 | ||
BC079149.1 | - | 9 | 861 | 7128541 |