Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024390.2 | NP_077366.2 | 6 | 660 | ||
GenBank mRNA | BC062399.1 | - | 6 | 660 | 5599288 | |
U44750.1 | - | 6 | 676 |