Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024397.2 | NP_077373.2 | 2, 3 | 353 | ||
XM_006254353.2 | XP_006254415.1 | 2, 3 | 424 | |||
XM_006254354.2 | XP_006254416.1 | 2, 3 | 423 | |||
XM_006254355.2 | XP_006254417.1 | 2, 3 | 423 | |||
XM_006254356.2 | XP_006254418.1 | 2, 3 | 423 | |||
XM_006254357.2 | XP_006254419.1 | 2, 3 | 423 | |||
XM_006254358.2 | XP_006254420.1 | 2, 3 | 423 | |||
XM_006254359.2 | XP_006254421.1 | 2, 3 | 423 | |||
XM_006254360.2 | XP_006254422.1 | 2, 3 | 441 | |||
GenBank mRNA | AF176784.1 | - | 2, 3 | 290 | ||
BC129092.1 | - | 2, 3 | 309 | 7192971 |