Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082539.1 | NP_001076008.1 | 2 | 652 | ||
NM_001082540.1 | NP_001076009.1 | 3 | 709 | |||
NM_001082541.1 | NP_001076010.1 | 2 | 652 | |||
NM_024404.2 | NP_077380.2 | 3 | 709 | |||
GenBank mRNA | AB046615.1 | - | 3 | 473 | ||
AB046616.1 | - | 2 | 367 | |||
AB046617.1 | - | 3 | 424 | |||
AB046618.1 | - | 2 | 367 | |||
CO560068.1 | - | 2 | 652 |