Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173103.1 | NP_775126.1 | 8 | 847 | ||
XM_006239320.2 | XP_006239382.1 | 11 | 3719 | |||
XM_008764293.1 | XP_008762515.1 | 10 | 3387 | |||
XM_008764294.1 | XP_008762516.1 | 10 | 1653 | |||
XM_008764295.1 | XP_008762517.1 | 9 | 1798 | |||
XM_008764296.1 | XP_008762518.1 | 9 | 1787 | |||
XM_008764297.1 | XP_008762519.1 | 8 | 1658 | |||
GenBank mRNA | BC081698.1 | - | 8 | 1342 | ||
D26111.1 | - | 9 | 901 | |||
DQ437671.1 | - | 9 | 951 | |||
DQ437672.1 | - | 9 | 844 | |||
DQ437673.1 | - | 8 | 822 | |||
DQ437674.1 | - | 8 | 786 | |||
DQ437675.1 | - | 9 | 794 | |||
DQ437676.1 | - | 8 | 772 | |||
Z30663.1 | - | 8 | 847 |