Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173103.1 | NP_775126.1 | 11 | 1216 | ||
XM_006239320.2 | XP_006239382.1 | 14 | 4088 | |||
XM_008764293.1 | XP_008762515.1 | 13 | 3756 | |||
XM_008764294.1 | XP_008762516.1 | 13 | 2022 | |||
XM_008764295.1 | XP_008762517.1 | 12 | 2167 | |||
XM_008764296.1 | XP_008762518.1 | 12 | 2156 | |||
XM_008764297.1 | XP_008762519.1 | 11 | 2027 | |||
GenBank mRNA | BC081698.1 | - | 11 | 1711 | ||
D26111.1 | - | 12 | 1270 | |||
DQ437671.1 | - | 12 | 1320 | |||
DQ437672.1 | - | 12 | 1213 | |||
DQ437673.1 | - | 11 | 1191 | |||
DQ437674.1 | - | 11 | 1155 | |||
DQ437675.1 | - | 12 | 1163 | |||
DQ437676.1 | - | 11 | 1141 | |||
Z30663.1 | - | 11 | 1216 |