Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 7 | 1030 | ||
XM_006716392.2 | XP_006716455.1 | 6 | 1119 | |||
GenBank mRNA | AF035277.1 | - | 7 | 878 | ||
AK090513.1 | - | 4 | 2694 | |||
AK315122.1 | - | 7 | 900 | |||
BC010550.1 | - | 7 | 831 | 4153601 | ||
BX647744.1 | - | 2 | 1851 | |||
CR456803.1 | - | 7 | 766 | |||
CR541850.1 | - | 7 | 766 | |||
KU178337.1 | - | 7 | 766 | |||
KU178338.1 | - | 5 | 487 | |||
S79669.1 | - | 7 | 918 | |||
U17280.1 | - | 7 | 892 |