Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 7 | 1321 | ||
XM_006716392.2 | XP_006716455.1 | 6 | 1410 | |||
GenBank mRNA | AF035277.1 | - | 7 | 1169 | ||
AK090513.1 | - | 4 | 2985 | |||
AY532319.1 | - | 1 | 287 | |||
BC010550.1 | - | 7 | 1122 | 4153601 | ||
BX647744.1 | - | 2 | 2142 | |||
U17280.1 | - | 7 | 1182 |