Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134760.1 | NP_001128232.1 | 3, 4 | 622 | ||
NM_032619.2 | NP_116008.2 | 3, 4 | 622 | |||
XM_006228691.2 | XP_006228753.1 | 3, 4 | 766 | |||
XM_006228692.2 | XP_006228754.1 | 3, 4 | 766 | |||
GenBank mRNA | BC169048.1 | - | 3, 4 | 577 |