Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 6 | 972 | ||
GenBank mRNA | AF035277.1 | - | 6 | 820 | ||
AK090513.1 | - | 4 | 1803 | |||
AK315122.1 | - | 6 | 842 | |||
BC010550.1 | - | 6 | 773 | 4153601 | ||
BX647744.1 | - | 2 | 958 | |||
CR456803.1 | - | 6 | 708 | |||
CR541850.1 | - | 6 | 708 | |||
KU178337.1 | - | 6 | 708 | |||
S79669.1 | - | 6 | 860 | |||
U17280.1 | - | 6 | 834 |